Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106052.1 | NP_001099522.1 | 3 - 4 | 589 | 108 | |
XM_008770863.1 | XP_008769085.1 | 3 - 4 | 1416 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Rattus norvegicus Probe spans exons |