Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc47a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC088413.1 | - | 7109897 | 135 | ||
FQ217802.1 | - | 135 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |