Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001055549.5 | XP_001055549.2 | 2 - 3 | 382 | 62 | |
XM_006257140.2 | XP_006257202.1 | 3 - 4 | 427 | 62 | ||
XM_008773329.1 | XP_008771551.1 | 2 - 3 | 2988 | 62 | ||
XM_008773330.1 | XP_008771552.1 | 2 - 3 | 2988 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |