Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brwd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773357.1 | XP_008771579.1 | 17 - 18 | 2102 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |