Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109451.1 | NP_001102921.1 | 307 | 124 |
Gene Symbol: LOC100910581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751155.2 | XP_003751203.1 | 1 - 2 | 320 | 124 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |