Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105953.2 | NP_001099423.2 | 3 - 4 | 234 | 64 | |
NM_001277263.1 | NP_001264192.1 | 3 - 4 | 311 | 64 | ||
GenBank mRNA | BC169086.1 | - | 2 - 3 | 201 | 5621176 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |