Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108708.1 | NP_001102178.1 | 3 - 4 | 425 | 73 | |
XM_006240022.2 | XP_006240084.1 | 3 - 4 | 892 | 73 | ||
XM_006240023.2 | XP_006240085.1 | 3 - 4 | 890 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |