Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015013.1 | NP_001015013.1 | 1 - 2 | 410 | 86 | |
GenBank mRNA | BC093377.1 | - | 1 - 2 | 410 | 7388027 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |