Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007658.1 | NP_001007659.1 | 1 - 2 | 603 | 120 | |
GenBank mRNA | BC083742.1 | - | 1 - 2 | 603 | 7191975 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |