Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108789.3 | NP_001102259.2 | 8 - 9 | 977 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |