Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106691.1 | NP_001100161.1 | 1 | 3337 | 125 | |
XM_006239150.2 | XP_006239212.1 | 4 | 4328 | 125 | ||
XM_006239152.2 | XP_006239214.1 | 3 | 4273 | 125 | ||
XM_008764217.1 | XP_008762439.1 | 3 | 3688 | 125 | ||
XM_008764218.1 | XP_008762440.1 | 4 | 3596 | 125 | ||
XM_008764219.1 | XP_008762441.1 | 3 | 3804 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |