Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025651.1 | NP_001020822.1 | 12 - 13 | 1347 | 77 | |
XM_006239013.2 | XP_006239075.1 | 12 - 13 | 2185 | 77 | ||
GenBank mRNA | BC098679.1 | - | 12 - 13 | 1347 | 7384784 | 77 |
FQ227771.1 | - | 3 - 4 | 323 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |