Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760837.1 | XP_008759059.1 | 8 - 9 | 2226 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |