Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013957.1 | NP_001013979.1 | 5 - 6 | 713 | 161 | |
XM_006256707.1 | XP_006256769.1 | 5 - 6 | 687 | 161 | ||
XM_006256708.2 | XP_006256770.1 | 4 - 5 | 588 | 161 | ||
GenBank mRNA | BC059163.1 | - | 5 - 6 | 713 | 6889015 | 161 |
FQ215363.1 | - | 2 - 3 | 299 | 161 | ||
FQ215670.1 | - | 2 - 3 | 297 | 161 | ||
FQ227645.1 | - | 2 - 3 | 300 | 161 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |