Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177689.1 | NP_001171160.1 | 1 - 2 | 46 | 70 | |
XM_008763336.1 | XP_008761558.1 | 2 - 3 | 2138 | 70 | ||
GenBank mRNA | FJ404473.1 | - | 1 - 2 | 82 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |