Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008876.2 | NP_001008876.1 | 3 - 4 | 202 | 132 | |
GenBank mRNA | AY310144.1 | - | 3 - 4 | 202 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |