Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klri2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012648.1 | NP_001012666.1 | 5 - 6 | 660 | 113 | |
XM_008763327.1 | XP_008761549.1 | 4 - 5 | 904 | 113 | ||
GenBank mRNA | AY324871.1 | - | 5 - 6 | 660 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |