Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013937.1 | NP_001013959.1 | 1 | 1331 | 81 | |
GenBank mRNA | BC083677.1 | - | 1 | 1331 | 7135827 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Both primers and probe map within a single exon |