Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024686.1 | NP_001019857.1 | 1 | 818 | 114 | |
GenBank mRNA | AY916510.1 | - | 1 | 818 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |