Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025135.1 | NP_001020306.1 | 1 | 172 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |