Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008971.1 | NP_001008971.1 | 1 | 38 | 71 | |
XM_006236848.1 | XP_006236910.1 | 3 | 374 | 71 | ||
GenBank mRNA | AY510293.1 | - | 1 | 38 | 71 | |
U36896.1 | - | 3 | 388 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |