Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
  • Special Offers
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Gene Expression
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Rn04223057_mH
            See other SULT2A1 GE Assays ›
            Gene Symbol:
            Sult2a1 LOC100360574 Sult2a6
            Gene Name:
            sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
            alcohol sulfotransferase-like
            sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 6
            Gene Aliases:
            STa, Smp-2, St2, St2a1, Sth2
            -
            RATSMP2A, RATSMP2B, Smp2a, St2a2, Sult2a4l1, Sult2al1
            Chromosome Location:
            Chr.1: 76558721 - 76614309 on Build Rnor_6.0
            Chromosome Location:
            Chr.1: 76004950 - 76053083 on Build Rnor_6.0
            Chromosome Location:
            Chr.1: 76529121 - 76722965 on Build Rnor_6.0
            Species:
            Rat
            Species Specific ID (Flybase ID):
            -
            -
            -
            Assay ID Rn04223057_mH
            Size
            Availability Inventoried
            Catalog # 4331182
            Price (USD) 272.00
            Your Price
            Online offer (USD):256.65
            256.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data...
            ×
            Back To Top

            Assay Details

            Gene Symbol: Sult2a1
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_131903.1 NP_571978.1 2 - 3 380 93
            GenBank mRNA D14987.1 - 2 - 3 380 93
            D14988.1 - 2 - 3 380 93
            FQ209430.1 - 2 - 3 255 93
            FQ209460.1 - 2 - 3 255 93
            FQ218232.1 - 2 - 3 255 93
            FQ219256.1 - 2 - 3 255 93
            FQ219272.1 - 2 - 3 276 93
            Gene Symbol: LOC100360574
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq XM_008759026.1 XP_008757248.1 2 - 3 343 93
            Gene Symbol: Sult2a6
            Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
            RefSeq NM_012695.3 NP_036827.3 2 - 3 380 93
            GenBank mRNA FQ209484.1 - 2 - 3 255 93
            FQ209563.1 - 2 - 3 255 93
            FQ218388.1 - 2 - 3 255 93
            FQ218712.1 - 2 - 3 255 93
            FQ219267.1 - 2 - 3 255 93
            FQ219345.1 - 2 - 3 255 93
            M33329.1 - 2 - 3 380 93
            X63410.1 - 2 - 3 380 93

            Back To Top

            More Information


            Updated Mapping Information

            This assay targets the same location as RTA transcript cluster(s) TC0100004843.rn.1

            Set Membership:

            Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus Inventoried Probe spans exons

            Matching Ambion siRNAs:

            Click here to find siRNAs for Gene Sult2a1
            Click here to find siRNAs for Gene LOC100360574
            Click here to find siRNAs for Gene Sult2a6

            Panther Classification:

            Molecular Function -

            transferase

            Gene Ontology Categories:

            Function(s) Process(es)

            aging
            metabolic process
            steroid metabolic process
            response to activity
            response to insecticide
            drug metabolic process
            response to nutrient levels
            sulfation
            cellular response to vitamin D
            biological_process
            alcohol sulfotransferase activity
            drug binding
            sulfotransferase activity
            transferase activity
            bile-salt sulfotransferase activity
            3'-phosphoadenosine 5'-phosphosulfate binding
            molecular_function

            MicroArray Probes:

            Agilent Affymetrix

            Array Name Probe IDs Targets Target Type
            Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P175204 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 180336 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 662215 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 399517 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 332724 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 696094 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 545895 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 546097 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 4823 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 91316 - -
            Agilent SurePrint G3 Rat CGH Microarray 1x1M 578331 - -
            Agilent SurePrint G3 Rat CGH Microarray 4x180K 142365 - -
            Agilent SurePrint G3 Rat CGH Microarray 4x180K 180487 - -
            Agilent SurePrint G3 Rat CGH Microarray 4x180K 52215 - -
            Agilent SurePrint G3 Rat CGH Microarray 4x180K 154160 - -
            Agilent SurePrint G3 Rat CGH Microarray 4x180K 169906 - -
            Agilent SurePrint G3 Rat CGH Microarray 4x180K 128799 - -
            Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175204 - -
            Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175204 - -
            Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P175204 - -
            SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P175204 - -
            SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13235 - -
            Agilent Rat Genome CGH Microarray 105A (G4436A) 75769 - -
            Agilent Rat Genome CGH Microarray 105A (G4436A) 28006 - -
            Array Name Probe IDs Targets Target Type
            Affymetrix Rat Gene 1.0 ST Array 10719191 - -
            Affymetrix Rat Gene 1.0 ST Array 10719188 - -
            Affymetrix Rat Gene 1.0 ST Array 10719190 - -
            Affymetrix Rat Gene 1.1 ST Array 10719187 - -
            Rat Genome 230 2.0 Array 1369570_at - -
            Rat Genome 230 2.0 Array 1369570_PM_at - -
            Affymetrix Rat Clariom S Assay PSR0100038182.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Clariom S Assay PSR0100038181.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Clariom S Assay PSR0100038187.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Clariom S Assay PSR0100038180.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Clariom S Assay PSR0100038183.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Clariom S Assay PSR0100038177.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Clariom S Assay PSR0100038178.rn.1 TC0100004843.rn.1 RTA TC
            Affymetrix Rat Gene 1.0 ST Array 10719187 - -
            Rat Genome U34 Set D14988_f_at - -
            Rat Genome U34 Set D14987_f_at - -
            Rat Genome U34 Set rc_AI028836_at - -
            Rat Genome U34 Set M31363mRNA_f_at - -
            Rat Genome U34 Set rc_AA945589_at - -
            Rat Genome U34 Set D14987_f_at - -
            Rat Genome U34 Set D14988_f_at - -
            Rat Genome U34 Set M31363mRNA_f_at - -
            Rat Genome U34 Set D14987_f_at - -
            Rat Genome U34 Set rc_AI028836_at - -
            Rat Genome U34 Set rc_AA945589_at - -

            Back To Top

            Related Products

            • TaqMan® Fast Advanced Master Mix
            • SuperScript® VILO™ cDNA Synthesis Kit
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-xpgds:80/100.66.76.20:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline