Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCGTGGTTTTGTTTTAAACCC[C/T]GAAGAAAATCCCCATGACAACATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614593 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf45 - chromosome 16 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA0430 - KIAA0430 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184998.1 | Intron | NP_001171927.1 | ||||
NM_001184999.1 | Intron | NP_001171928.1 | ||||
NM_014647.3 | Intron | NP_055462.2 | ||||
XM_005255704.1 | Intron | XP_005255761.1 | ||||
XM_005255706.1 | Intron | XP_005255763.1 | ||||
XM_005255707.1 | Intron | XP_005255764.1 | ||||
XM_017023900.1 | Intron | XP_016879389.1 | ||||
XM_017023901.1 | Intron | XP_016879390.1 | ||||
XM_017023902.1 | Intron | XP_016879391.1 | ||||
XM_017023903.1 | Intron | XP_016879392.1 | ||||
XM_017023904.1 | Intron | XP_016879393.1 | ||||
XM_017023905.1 | Intron | XP_016879394.1 | ||||
XM_017023906.1 | Intron | XP_016879395.1 |