Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAAAAACTCCTATTTATCATGG[A/C]TGTGCCAGGATCGAGAGAATCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYSL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPYSL3 - dihydropyrimidinase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197294.1 | 3303 | UTR 3 | NP_001184223.1 | |||
NM_001387.2 | 3303 | UTR 3 | NP_001378.1 | |||
XM_011537574.1 | 3303 | UTR 3 | XP_011535876.1 |
STK32A - serine/threonine kinase 32A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112724.1 | 3303 | Intron | NP_001106195.1 | |||
NM_001287740.1 | 3303 | Intron | NP_001274669.1 | |||
NM_145001.3 | 3303 | Intron | NP_659438.1 | |||
XM_011537576.1 | 3303 | Intron | XP_011535878.1 | |||
XM_011537577.2 | 3303 | Intron | XP_011535879.1 | |||
XM_011537579.2 | 3303 | Intron | XP_011535881.1 | |||
XM_017009213.1 | 3303 | Intron | XP_016864702.1 | |||
XM_017009214.1 | 3303 | Intron | XP_016864703.1 |