Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATTTGCCTTATGCAGCTACCTG[C/T]TTATTGCAGTGAAGGGTACTGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDK1 - pyruvate dehydrogenase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278549.1 | Intron | NP_001265478.1 | ||||
NM_002610.4 | Intron | NP_002601.1 | ||||
XM_006712594.2 | Intron | XP_006712657.1 | ||||
XM_006712595.3 | Intron | XP_006712658.1 | ||||
XM_011511343.2 | Intron | XP_011509645.1 | ||||
XM_011511344.2 | Intron | XP_011509646.1 | ||||
XM_011511345.2 | Intron | XP_011509647.1 | ||||
XM_011511347.2 | Intron | XP_011509649.1 |