Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGTTCTCCTGGGCCCTCCGGAT[C/T]AGGTAGGGGATTACCTCCTCCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01529 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01529 - long intergenic non-protein coding RNA 1529 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRODH2 - proline dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021232.1 | 1369 | Silent Mutation | CTA,CTG | L,L 498 | NP_067055.1 | |
XM_017027084.1 | 1369 | Silent Mutation | CTA,CTG | L,L 422 | XP_016882573.1 | |
XM_017027085.1 | 1369 | UTR 3 | XP_016882574.1 |