Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCACATTATTTGCTTCTTCCAT[A/C]CCATTCTTCATATTGATCATTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF44 - zinc finger protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164276.1 | Intron | NP_001157748.1 | ||||
NM_016264.3 | Intron | NP_057348.3 | ||||
XM_005259934.3 | Intron | XP_005259991.1 | ||||
XM_011528062.2 | Intron | XP_011526364.1 | ||||
XM_011528064.1 | Intron | XP_011526366.1 | ||||
XM_011528066.1 | Intron | XP_011526368.1 | ||||
XM_017026870.1 | Intron | XP_016882359.1 | ||||
XM_017026871.1 | Intron | XP_016882360.1 |