Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTCCCAGCTGTGGAGCAGGAGG[A/G]GGGAGGGTTAGGGTCTCCTCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604637 MIM: 138450 MIM: 615521 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA4L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFA4L2 - NDUFA4, mitochondrial complex associated like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020142.3 | Intron | NP_064527.1 | ||||
XM_005269033.2 | Intron | XP_005269090.1 | ||||
XM_011538573.2 | Intron | XP_011536875.1 |
NXPH4 - neurexophilin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHMT2 - serine hydroxymethyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166356.1 | Intron | NP_001159828.1 | ||||
NM_001166357.1 | Intron | NP_001159829.1 | ||||
NM_001166358.1 | Intron | NP_001159830.1 | ||||
NM_001166359.1 | Intron | NP_001159831.1 | ||||
NM_005412.5 | Intron | NP_005403.2 | ||||
XM_011538675.2 | Intron | XP_011536977.1 | ||||
XM_011538676.2 | Intron | XP_011536978.1 | ||||
XM_011538677.2 | Intron | XP_011536979.1 | ||||
XM_011538678.2 | Intron | XP_011536980.1 |
STAC3 - SH3 and cysteine rich domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |