Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAATTCAGGAGCCCAACTGCAT[A/G]TTACCTACTTGGTGTCTGGCAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611415 | ||||||||||||||||||||
Literature Links: |
POLD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLD3 - DNA polymerase delta 3, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006591.2 | 304 | Missense Mutation | ATT,GTT | I,V 59 | NP_006582.1 | |
XM_005273716.1 | 304 | Missense Mutation | ATT,GTT | I,V 59 | XP_005273773.1 | |
XM_011544734.2 | 304 | Missense Mutation | ATT,GTT | I,V 41 | XP_011543036.1 |