Up to 50% off benchtop instruments, accessories, consumables and more

Save now

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

SNP Genotyping
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › C_163144269_10
            See other INSM2 GT Assays ›
            SNP ID:
            rs146562217
            Gene
            INSM2 RALGAPA1
            Gene Name
            INSM transcriptional repressor 2
            Ral GTPase activating protein catalytic alpha subunit 1
            Set Membership:
            -
            Chromosome Location:
            Chr.14: 35539573 - 35539573 on Build GRCh38
            Polymorphism:
            C/T, Transition substitution
            Context Sequence [VIC/FAM]:

            TTTATTGGCTGAGCCAGAGGAACGA[C/T]GCAGCTTCATGGACATGCGGCTTTT

            Assay ID C_163144269_10
            Size
            Availability Made To Order
            Catalog # 4351379
            Price (USD) 442.00
            Your Price
            Online offer (USD):407.65
            407.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details



            Species:

            Human

            dbSNP Submissions:

            NA

            Phenotype:

            MIM: 614027 MIM: 608884

            Literature Links:

            INSM2 PubMed Links

            Allele Nomenclature:

            Minor Allele Frequency:

            1000Genome Applied Biosystems® HapMap
            Global
            T (0.00)
            (1.00)
            Caucasian - Not Available CEPH (CEU) - Not Available
            EAS
            T (0.00)
            (1.00)
            African American - Not Available YRI (Yoruba) - Not Available
            SAS
            T (0.01)
            (0.99)
            Chinese - Not Available CHB (Han Chinese) - Not Available
            AFR
            T (0.00)
            (1.00)
            Japanese - Not Available JPT (Japanese) - Not Available
            EUR
            T (0.00)
            (1.00)
            AMR
            T (0.00)
            (1.00)
            INSM2 - INSM transcriptional repressor 2
            There are no transcripts associated with this gene.
            RALGAPA1 - Ral GTPase activating protein catalytic alpha subunit 1
            Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
            NM_001283043.1 5880 Intron NP_001269972.1
            NM_001283044.1 5880 Intron NP_001269973.1
            NM_014990.1 5880 Intron NP_055805.1
            NM_194301.2 5880 Intron NP_919277.2
            XM_005267491.3 5880 Intron XP_005267548.1
            XM_005267492.3 5880 Intron XP_005267549.1
            XM_005267493.3 5880 Intron XP_005267550.1
            XM_005267498.4 5880 Missense Mutation CAT,CGT H,R 1316 XP_005267555.1
            XM_006720098.2 5880 Intron XP_006720161.1
            XM_006720099.2 5880 Intron XP_006720162.1
            XM_006720100.3 5880 Intron XP_006720163.1
            XM_006720101.2 5880 Intron XP_006720164.1
            XM_006720102.3 5880 Intron XP_006720165.1
            XM_006720103.2 5880 Intron XP_006720166.1
            XM_006720104.3 5880 Intron XP_006720167.1
            XM_011536615.2 5880 Intron XP_011534917.1
            XM_011536616.2 5880 Intron XP_011534918.1
            XM_017021141.1 5880 Intron XP_016876630.1
            XM_017021142.1 5880 Intron XP_016876631.1
            XM_017021143.1 5880 Intron XP_016876632.1
            XM_017021144.1 5880 Intron XP_016876633.1
            XM_017021145.1 5880 Intron XP_016876634.1
            XM_017021146.1 5880 Missense Mutation CAT,CGT H,R 1825 XP_016876635.1
            XM_017021147.1 5880 UTR 3 XP_016876636.1

            Back To Top

            More Information


            Panther Classification:

            Molecular Function -

            GTPase-activating protein

            Gene Ontology Categories:

            Function(s) Process(es)

            regulation of transcription, DNA-templated
            regulation of small GTPase mediated signal transduction
            activation of GTPase activity
            GTPase activator activity
            protein heterodimerization activity

            Back To Top

            Related Products

            • TaqMan® Genotyping Master Mix
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-sbk6v:80/100.66.72.122:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline