Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTATACCCGAAGACGTACTGG[A/G]GGCGCTCGGCGGCCCGTTTGAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608243 | ||||||||||||||||||||
Literature Links: |
FAM189A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM189A1 - family with sequence similarity 189 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015307.1 | 512 | Intron | NP_056122.1 | |||
XM_011521407.2 | 512 | Intron | XP_011519709.1 |
NSMCE3 - NSE3 homolog, SMC5-SMC6 complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138704.3 | 512 | Missense Mutation | CCC,CTC | P,L 134 | NP_619649.1 |