Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTGTACCAAATCTCCCAAAGC[A/G]TGTACAAATGCAGCTCTCACTGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602095 | ||||||||||||||||||||
Literature Links: |
SLC30A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC30A4 - solute carrier family 30 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321036.1 | 949 | Intron | NP_001307965.1 | |||
NM_013309.5 | 949 | Silent Mutation | CAC,CAT | H,H 277 | NP_037441.2 | |
XM_011521997.2 | 949 | Silent Mutation | CAC,CAT | H,H 277 | XP_011520299.1 | |
XM_017022560.1 | 949 | Silent Mutation | CAC,CAT | H,H 277 | XP_016878049.1 | |
XM_017022561.1 | 949 | Intron | XP_016878050.1 | |||
XM_017022562.1 | 949 | Intron | XP_016878051.1 |