Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTTCAGGCTGTACAGGTAGGAG[A/C]CCTTGTGCGGCGAGCCAAAGTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611026 | ||||||||||||||||||||
Literature Links: |
FA2H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FA2H - fatty acid 2-hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024306.4 | 752 | Missense Mutation | GGC,GTC | G,V 326 | NP_077282.3 | |
XM_011523317.2 | 752 | Intron | XP_011521619.1 | |||
XM_011523319.2 | 752 | Missense Mutation | GGC,GTC | G,V 246 | XP_011521621.1 |