Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTCCTCACCTCCCCTCGAATC[A/G]GGTCTGGGCTGCCCACCACGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614357 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSM1 - acyl-CoA synthetase medium-chain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318890.1 | 2409 | Missense Mutation | NP_001305819.1 | |||
NM_052956.2 | 2409 | Missense Mutation | NP_443188.2 | |||
XM_006721016.2 | 2409 | Missense Mutation | XP_006721079.1 | |||
XM_006721017.2 | 2409 | Missense Mutation | XP_006721080.1 | |||
XM_006721018.2 | 2409 | Missense Mutation | XP_006721081.1 | |||
XM_011545729.2 | 2409 | Intron | XP_011544031.1 | |||
XM_011545730.2 | 2409 | Intron | XP_011544032.1 | |||
XM_011545731.2 | 2409 | Missense Mutation | XP_011544033.1 | |||
XM_011545732.2 | 2409 | Missense Mutation | XP_011544034.1 | |||
XM_017022914.1 | 2409 | Intron | XP_016878403.1 | |||
XM_017022915.1 | 2409 | Missense Mutation | XP_016878404.1 |