Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCGCAGCAGGACCAGGGCCACC[C/T]GGAAGATGATCTTAACGCCTGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604470 MIM: 613620 | ||||||||||||||||||||
Literature Links: |
CD2BP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD2BP2 - CD2 cytoplasmic tail binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10B - TBC1 domain family member 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015527.3 | 1546 | Missense Mutation | CAG,CGG | Q,R 554 | NP_056342.3 | |
XM_011545789.1 | 1546 | Missense Mutation | CAG,CGG | Q,R 449 | XP_011544091.1 | |
XM_011545790.2 | 1546 | Missense Mutation | AGG,GGG | R,G 491 | XP_011544092.1 |