Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTCCAGCTGAGTGAAGCTAAG[A/G]TATTTTCTGAGCCAACAAAGATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TK2 - thymidine kinase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172643.1 | 2010 | UTR 3 | NP_001166114.1 | |||
NM_001172644.1 | 2010 | UTR 3 | NP_001166115.1 | |||
NM_001172645.1 | 2010 | UTR 3 | NP_001166116.1 | |||
NM_001271934.1 | 2010 | UTR 3 | NP_001258863.1 | |||
NM_001271935.1 | 2010 | UTR 3 | NP_001258864.1 | |||
NM_001272050.1 | 2010 | UTR 3 | NP_001258979.1 | |||
NM_004614.4 | 2010 | UTR 3 | NP_004605.4 |