Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGACGAACTGGACTCCAGCCAG[C/T]TCCAGATGGAGTCCGATGAGGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602384 MIM: 600307 | ||||||||||||||||||||
Literature Links: |
LOC107984973 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107984973 - atherin-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017025490.1 | 133 | Intron | XP_016880979.1 |
PLD2 - phospholipase D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243108.1 | 133 | Missense Mutation | CTC,TTC | L,F 20 | NP_001230037.1 | |
NM_002663.4 | 133 | Missense Mutation | CTC,TTC | L,F 20 | NP_002654.3 | |
XM_005256696.2 | 133 | Intron | XP_005256753.1 | |||
XM_017024764.1 | 133 | UTR 5 | XP_016880253.1 |
PSMB6 - proteasome subunit beta 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |