Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCCAACCTCGCCAACTTCGCTTA[C/T]GACCCCAGCAACTACGAGTATCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603879 | ||||||||||||||||||||
Literature Links: |
ARMC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC7 - armadillo repeat containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304271.1 | 472 | Silent Mutation | TAC,TAT | Y,Y 45 | NP_001291200.1 | |
NM_001304272.1 | 472 | Silent Mutation | TAC,TAT | Y,Y 45 | NP_001291201.1 | |
NM_001304273.1 | 472 | Silent Mutation | TAC,TAT | Y,Y 45 | NP_001291202.1 | |
NM_024585.3 | 472 | Silent Mutation | TAC,TAT | Y,Y 45 | NP_078861.1 |
SLC16A5 - solute carrier family 16 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |