Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTCTCATTTTTCAAATCATTC[C/T]TCTGCACTCACTTTCTATAATGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF546 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF546 - zinc finger protein 546 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297763.1 | 318 | Intron | NP_001284692.1 | |||
NM_178544.4 | 318 | Missense Mutation | CCT,CTT | P,L 21 | NP_848639.2 | |
XM_011526898.2 | 318 | Intron | XP_011525200.1 | |||
XM_011526899.2 | 318 | Missense Mutation | CCT,CTT | P,L 21 | XP_011525201.1 |