Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACGAGGTCCAGGTTGGTTCGGA[C/T]TTGAACAGCTCGGGACCATTGATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610349 MIM: 609623 | ||||||||||||||||||||
Literature Links: |
MAMSTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAMSTR - MEF2 activating motif and SAP domain containing transcriptional regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASIP1 - Ras interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017805.2 | 2436 | Missense Mutation | ATC,GTC | I,V 805 | NP_060275.2 | |
XM_011527053.2 | 2436 | Missense Mutation | ATC,GTC | I,V 906 | XP_011525355.2 | |
XM_011527054.2 | 2436 | Missense Mutation | ATC,GTC | I,V 906 | XP_011525356.2 | |
XM_011527055.2 | 2436 | Missense Mutation | ATC,GTC | I,V 808 | XP_011525357.2 | |
XM_017026914.1 | 2436 | Missense Mutation | ATC,GTC | I,V 906 | XP_016882403.1 | |
XM_017026915.1 | 2436 | Missense Mutation | ATC,GTC | I,V 808 | XP_016882404.1 |