Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGGACTGGAAGACATTAGAAT[A/C]AGGTTTGTCATTGAGTGTGTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAO2 - hydroxyacid oxidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005783.2 | 125 | UTR 5 | NP_001005783.2 | |||
NM_016527.3 | 125 | UTR 5 | NP_057611.1 | |||
XM_011541558.1 | 125 | Intron | XP_011539860.1 | |||
XM_011541559.1 | 125 | Intron | XP_011539861.1 | |||
XM_011541560.1 | 125 | Intron | XP_011539862.1 | |||
XM_011541561.1 | 125 | Intron | XP_011539863.1 | |||
XM_017001421.1 | 125 | Intron | XP_016856910.1 |
HAO2-IT1 - HAO2 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |