Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAACCTTACTGGCAGATCCTTC[A/G]CTGTTCTGTCTTTGCAGTGCTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608069 | ||||||||||||||||||||
Literature Links: |
ERRFI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERRFI1 - ERBB receptor feedback inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018948.3 | 1370 | Silent Mutation | AGC,AGT | S,S 369 | NP_061821.1 | |
XM_005263477.2 | 1370 | Silent Mutation | AGC,AGT | S,S 318 | XP_005263534.1 | |
XM_006710697.2 | 1370 | Silent Mutation | AGC,AGT | S,S 369 | XP_006710760.1 | |
XM_011541596.2 | 1370 | Silent Mutation | AGC,AGT | S,S 294 | XP_011539898.1 |