Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATGCGCCACGAGTCACATGAT[A/G]TCCACGAAGAACTCGCAGGGGTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615467 MIM: 601365 MIM: 605865 | ||||||||||||||||||||
Literature Links: |
CPTP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPTP - ceramide-1-phosphate transfer protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DVL1 - dishevelled segment polarity protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004421.2 | 2156 | Silent Mutation | GAC,GAT | D,D 668 | NP_004412.2 | |
XM_005244731.3 | 2156 | Silent Mutation | GAC,GAT | D,D 693 | XP_005244788.1 | |
XM_005244732.3 | 2156 | Intron | XP_005244789.1 | |||
XM_005244733.3 | 2156 | Intron | XP_005244790.1 |
MIR6808 - microRNA 6808 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAS1R3 - taste 1 receptor member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152228.2 | 2156 | Intron | NP_689414.1 | |||
XM_017002435.1 | 2156 | Intron | XP_016857924.1 | |||
XM_017002436.1 | 2156 | Intron | XP_016857925.1 |