Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCATAACAAAGTTCGGCTGAC[A/G]GCAACAGACAAAACATTCCCGAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VAV3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VAV3 - vav guanine nucleotide exchange factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079874.1 | 3236 | UTR 3 | NP_001073343.1 | |||
NM_006113.4 | 3236 | UTR 3 | NP_006104.4 | |||
XM_005270360.2 | 3236 | UTR 3 | XP_005270417.1 | |||
XM_005270361.1 | 3236 | Intron | XP_005270418.1 | |||
XM_017000053.1 | 3236 | UTR 3 | XP_016855542.1 | |||
XM_017000054.1 | 3236 | Intron | XP_016855543.1 | |||
XM_017000055.1 | 3236 | UTR 3 | XP_016855544.1 | |||
XM_017000056.1 | 3236 | UTR 3 | XP_016855545.1 |