Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAAGCAGAAGCTGGCGTCGGCT[C/G]TTTTGGCTGAGCAGGAACCGCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603503 MIM: 609277 | ||||||||||||||||||||
Literature Links: |
DPM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPM1 - dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317034.1 | 123 | Intron | NP_001303963.1 | |||
NM_001317035.1 | 123 | Intron | NP_001303964.1 | |||
NM_001317036.1 | 123 | Intron | NP_001303965.1 | |||
NM_003859.2 | 123 | Intron | NP_003850.1 | |||
XM_011529093.2 | 123 | Intron | XP_011527395.1 |
MOCS3 - molybdenum cofactor synthesis 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014484.4 | 123 | Missense Mutation | CTT,GTT | L,V 32 | NP_055299.1 |