Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGGCCTGCATCCCTCACCTCTG[C/T]TTCCCAGTCTTCTGCTGACCTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MORC2 - MORC family CW-type zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303256.2 | 4795 | UTR 3 | NP_001290185.1 | |||
NM_001303257.2 | 4795 | UTR 3 | NP_001290186.1 | |||
NM_014941.3 | 4795 | UTR 3 | NP_055756.1 | |||
XM_011530004.1 | 4795 | Intron | XP_011528306.1 | |||
XM_017028667.1 | 4795 | UTR 3 | XP_016884156.1 | |||
XM_017028668.1 | 4795 | Intron | XP_016884157.1 |
MORC2-AS1 - MORC2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |