Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCGTCTCCGCAGCGCGCCCAACT[G/T]CATCCTCCTGGCCATGTTCCTCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610916 MIM: 184753 | ||||||||||||||||||||
Literature Links: |
NSUN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSUN2 - NOP2/Sun RNA methyltransferase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193455.1 | 509 | Intron | NP_001180384.1 | |||
NM_017755.5 | 509 | Intron | NP_060225.4 |
SRD5A1 - steroid 5 alpha-reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001047.3 | 509 | Missense Mutation | TGC,TTC | C,F 85 | NP_001038.1 | |
NM_001324322.1 | 509 | Missense Mutation | GCA,TCA | A,S 94 | NP_001311251.1 | |
NM_001324323.1 | 509 | UTR 5 | NP_001311252.1 |