Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTCGTTTGCCAGATATATTCGT[C/T]GGAATATAAGAGCTGCCAATGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616175 | ||||||||||||||||||||
Literature Links: |
UBE2J1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2J1 - ubiquitin conjugating enzyme E2 J1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016021.2 | 1242 | Missense Mutation | CAA,CGA | Q,R 304 | NP_057105.2 | |
XM_011535887.1 | 1242 | Missense Mutation | CAA,CGA | Q,R 264 | XP_011534189.1 | |
XM_011535888.2 | 1242 | UTR 3 | XP_011534190.1 |