Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGCGCAGCCAACGCTTCCTCA[A/G]TGATCCCGGGCATTTACTCTGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612524 MIM: 605700 | ||||||||||||||||||||
Literature Links: |
RPP21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPP21 - ribonuclease P/MRP subunit p21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199120.1 | 1362 | Missense Mutation | AAT,AGT | N,S 111 | NP_001186049.1 | |
NM_001199121.1 | 1362 | Missense Mutation | AAT,AGT | N,S 103 | NP_001186050.1 | |
NM_024839.2 | 1362 | Missense Mutation | AAT,AGT | N,S 103 | NP_079115.1 |
TRIM39 - tripartite motif containing 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM39-RPP21 - TRIM39-RPP21 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199119.1 | 1362 | Missense Mutation | AAT,AGT | N,S 452 | NP_001186048.1 |