Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCAGCACGGCCGCGCGCCCGA[A/T]CTGGTCGTTGCGTCGCTTCTCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM84B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM84B - family with sequence similarity 84 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174911.4 | 1307 | Missense Mutation | ATC,TTC | I,F 260 | NP_777571.1 | |
XM_017013107.1 | 1307 | Missense Mutation | ATC,TTC | I,F 260 | XP_016868596.1 | |
XM_017013108.1 | 1307 | Missense Mutation | ATC,TTC | I,F 242 | XP_016868597.1 |